Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A. aeolicus BPL/BioID2 Antibody (SS 3A5-E2), HRP, Novus Biologicals™
SDP

Catalog No. NBP259941H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-59941H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-59941H Supplier Novus Biologicals Supplier No. NBP259941H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A. aeolicus BPL/BioID2 Monoclonal specifically detects A. aeolicus BPL/BioID2 in Bacteria samples. It is validated for Western Blot.

Specifications

Antigen A. aeolicus BPL/BioID2
Applications Western Blot
Classification Monoclonal
Clone SS 3A5-E2
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Host Species Mouse
Immunogen GST fused to A. aeolicus BPL/BioID2
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Epitope Tags
Primary or Secondary Primary
Test Specificity This antibody is reactive with A. aeolicus and has not been tested with other species. The birA Entrez Gene number is NP_213397. UniProt number is O66837.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.