Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A. aeolicus BPL/BioID2 Antibody (SS QD1), PE, Novus Biologicals™
SDP

Catalog No. NB308755 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308755 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308755 Supplier Novus Biologicals Supplier No. NBP259940PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A. aeolicus BPL/BioID2 Monoclonal antibody specifically detects A. aeolicus BPL/BioID2 in Bacteria samples. It is validated for Western Blot, Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen A. aeolicus BPL/BioID2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone SS QD1
Conjugate PE
Dilution Western Blot, Simple Western, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Host Species Mouse
Immunogen GST fused to A. aeolicus BPL/BioID2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.