Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A-RAF Rabbit anti-Human, Clone: 4U9L7, Novus Biologicals™
SDP

Catalog No. NB168446 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168446 20 μg
NB168445 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168446 Supplier Novus Biologicals Supplier No. NBP31667720UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

A-RAF Monoclonal antibody specifically detects A-RAF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen A-RAF
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4U9L7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:10-1:20
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ARAF1A-RAF, EC 2.7.11, EC 2.7.11.1, Oncogene ARAF1, PKS, PKS2A-Raf proto-oncogene serine/threonine-protein kinase, Proto-oncogene A-Raf, Proto-oncogene A-Raf-1, Proto-oncogene Pks, RAFA1, Ras-binding protein DA-Raf, serine/threonine-protein kinase A-Raf, v-raf murine sarcoma 3611 viral oncogene homolog, v-raf murine sarcoma 3611 viral oncogene homolog 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 166-290 of human A-RAF (P10398).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 369
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.