Learn More
A20/TNFAIP3 Antibody (59A426) - BSA Free, Novus Biologicals™
SDP
Mouse Monoclonal Antibody has been used in 41 publication
Supplier:  Novus Biologicals NBP177533SS
 View more versions of this product
Catalog No. NBP177533SS
Only null left
Add to Cart
Description
Description
A20/TNFAIP3 Monoclonal specifically detects A20/TNFAIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.
Specifications
Specifications
A20/TNFAIP3
Monoclonal
1.0 mg/mL
Western Blot 2-4 ug/ml, Simple Western 1:100, Flow Cytometry 2.5 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml. Use reported in scientific literature (Metellus (2010)), Flow (Intracellular) 2.5 ug/ml. Use reported in scientific literature (Hjelmeland (2010)), CyTOF-ready
A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20
Mouse
Protein G purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
59A426
Unconjugated
P21580
TNFAIP3
Full length recombinant human A20/TNFAIP3 Antibody (59A426). The epitope has been mapped to the C-terminal portion of A20 (amino acids 440-790)
0.025 mg
Cancer, Necroptosis, Signal Transduction
7128
Store at -20C. Avoid freeze-thaw cycles.
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only