Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A20/TNFAIP3 Antibody (59A426) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 41 publication

$207.00 - $596.50

Specifications

Antigen A20/TNFAIP3
Clone 59A426
Concentration 1.0 mg/mL
Dilution Western Blot 2-4 ug/ml, Simple Western 1:100, Flow Cytometry 2.5 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml. Use reported in scientific literature (Metellus (2010)), Flow (Intracellular) 2.5 ug/ml. Use reported in scientific literature (Hjelmeland (2010)), CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP177533SS
SDP
View Documents
Novus Biologicals
NBP177533SS
0.025 mg
Each for $207.00
Only null left
Add to Cart
 
NBP177533
SDP
View Documents
Novus Biologicals
NBP177533
0.1 mg
Each for $596.50
Only null left
Add to Cart
 
Description

Description

A20/TNFAIP3 Monoclonal specifically detects A20/TNFAIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.
Specifications

Specifications

A20/TNFAIP3
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
Unconjugated
Mouse
Cancer, Necroptosis, Signal Transduction
P21580
7128
Full length recombinant human A20/TNFAIP3 Antibody (59A426). The epitope has been mapped to the C-terminal portion of A20 (amino acids 440-790)
Primary
Store at -20C. Avoid freeze-thaw cycles.
59A426
Western Blot 2-4 ug/ml, Simple Western 1:100, Flow Cytometry 2.5 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml. Use reported in scientific literature (Metellus (2010)), Flow (Intracellular) 2.5 ug/ml. Use reported in scientific literature (Hjelmeland (2010)), CyTOF-ready
Monoclonal
Purified
RUO
Human, Mouse, Rat
A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20
TNFAIP3
IgG1 κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.