Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A20/TNFAIP3 Antibody (59A426), FITC, Novus Biologicals™
SDP

Catalog No. NBP177533F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177533F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177533F Supplier Novus Biologicals Supplier No. NBP177533F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A20/TNFAIP3 Monoclonal specifically detects A20/TNFAIP3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen A20/TNFAIP3
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 59A426
Conjugate FITC
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20
Gene Symbols TNFAIP3
Host Species Mouse
Immunogen Full length recombinant human A20/TNFAIP3 Antibody (59A426). The epitope has been mapped to the C-terminal portion of A20 (amino acids 440-790)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Necroptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7128
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.