Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A20/TNFAIP3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220671 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220671 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220671 Supplier Novus Biologicals Supplier No. NBP220671
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A20/TNFAIP3 Polyclonal specifically detects A20/TNFAIP3 in Human samples. It is validated for Western Blot.

Specifications

Antigen A20/TNFAIP3
Applications Western Blot
Classification Polyclonal
Concentration 0.63mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P21580
Gene Alias A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20
Gene Symbols TNFAIP3
Host Species Rabbit
Immunogen Recombinant Protein encompassing a sequence within the C-terminus region of human A20/TNFAIP3 Antibody. The exact sequence is proprietary.
Molecular Weight of Antigen 90 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7128
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.