Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A20/TNFAIP3 Antibody (TNFAIP3/2813), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB391083 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391083 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391083 Supplier Novus Biologicals Supplier No. NBP308733AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A20/TNFAIP3 Monoclonal antibody specifically detects A20/TNFAIP3 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen A20/TNFAIP3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TNFAIP3/2813
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20
Host Species Mouse
Immunogen Recombinant fragment (around aa 241-356) of human A20/TNFAIP3 protein (exact sequence is proprietary) (Uniprot: P21580)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Inflammation, Innate Immunity, Necroptosis, Neurodegeneration, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7128
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.