Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A2BP1 Antibody (OTI4F9), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP271965R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271965R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271965R Supplier Novus Biologicals Supplier No. NBP271965R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A2BP1 Monoclonal specifically detects A2BP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen A2BP1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4F9
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2BP1, A2BP, A2BP1 Hexaribonucleotide-binding protein 1, Ataxin-2-binding protein 1, FOX-1, FOX1 hexaribonucleotide binding protein 1, Fox-1 homolog A, HRNBP1 ataxin 2-binding protein 1, RNA binding protein fox-1 homolog 1, RNA binding protein, fox-1 homolog (C. elegans) 1
Gene Symbols RBFOX1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-210 of human A2BP1 (NP_061193) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Epigenetics, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54715.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.