Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A2BP1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB335909 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB335909 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB335909 Supplier Novus Biologicals Supplier No. NBP298015PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A2BP1 Polyclonal antibody specifically detects A2BP1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen A2BP1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 2BP1, A2BP, A2BP1 Hexaribonucleotide-binding protein 1, Ataxin-2-binding protein 1, FOX-1, FOX1 hexaribonucleotide binding protein 1, Fox-1 homolog A, HRNBP1 ataxin 2-binding protein 1, RNA binding protein fox-1 homolog 1, RNA binding protein, fox-1 homolog (C. elegans) 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human A2BP1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54715
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.