Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A33/GPA33 Antibody (071), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB335187 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB335187 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB335187 Supplier Novus Biologicals Supplier No. NBP289873MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

A33/GPA33 Monoclonal antibody specifically detects A33/GPA33 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen A33/GPA33
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 071
Conjugate mFluor Violet 500 SE
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias A33, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), MGC129986, MGC129987, transmembrane glycoprotein A33
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human A33/GPA33 (Uniprot#: Q99795-1, Met1-Val235)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10223
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.