Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A4GNT Antibody (OTI2H9), HRP, Novus Biologicals™
SDP

Catalog No. NBP271905H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271905H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271905H Supplier Novus Biologicals Supplier No. NBP271905H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A4GNT Monoclonal specifically detects A4GNT in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen A4GNT
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2H9
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias alpha-1,4-N-acetylglucosaminyltransferase
Gene Symbols A4GNT
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 121-340 of human A4GNT (NP_057245) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 51146.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.