Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A4GNT Antibody (OTI2H9), PE, Novus Biologicals™
SDP

Catalog No. NB288583 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288583 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288583 Supplier Novus Biologicals Supplier No. NBP271905PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A4GNT Monoclonal antibody specifically detects A4GNT in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen A4GNT
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2H9
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias alpha-1,4-N-acetylglucosaminyltransferase
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 121-340 of human A4GNT (NP_057245) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51146
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.