Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ A66

Click to view available options
Quantity:
10 mg
50 mg
Description
A66 is a potent and selective PI 3-kinase p110α inhibitor (IC50 = 32 nM). Exhibits >100-fold selectivity for p110α over other PI 3-kinase isoforms. Inhibits Akt signaling and tumor growth in SK-OV-3 xenografts in mice.
Specifications
Specifications
Chemical Name or Material | (2S)-N1-[2-(1,1-Dimethylethyl)-4′-methyl[4,5′-bithiazol]-2′-yl]-1,2-pyrrolidinedicarboxamide |
CAS | 1166227-08-2 |
Quantity | 10 mg |
Target | PI 3-Kinase Inhibitors |
Molecular Formula | C17H23N5O2S2 |
Formula Weight | 393.53 |
Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction