Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A66
GREENER_CHOICE

Catalog No. p-7110964
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 559550

Supplier: Tocris Bioscience™ 5595/50

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective PI 3-kinase p110α inhibitor

A66 is a potent and selective PI 3-kinase p110α inhibitor (IC50 = 32 nM). Exhibits >100-fold selectivity for p110α over other PI 3-kinase isoforms. Inhibits Akt signaling and tumor growth in SK-OV-3 xenografts in mice.

Specifications

Chemical Name or Material (2S)-N1-[2-(1,1-Dimethylethyl)-4′-methyl[4,5′-bithiazol]-2′-yl]-1,2-pyrrolidinedicarboxamide
CAS 1166227-08-2
Quantity 50 mg
Target PI 3-Kinase Inhibitors
Molecular Formula C17H23N5O2S2
Formula Weight 393.53
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.