Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AA 147
GREENER_CHOICE

Catalog No. p-7161592
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

ER proteostasis regulator

AA 147 is an endoplasmic reticulum (ER) proteostasis regulator. Activates the ATF6 transcriptional program independently of ER stress and/or global UPR activation. Selectively reduces secretion of amyloidogenic transthyretin (TTR) from HepG2 cells. Reduces amyloidogenic immunoglobulin light chain (ALLC) secretion from ALMC-2 cells (EC50 = 1.1 μM).

Specifications

Chemical Name or Material N-(2-Hydroxy-5-methylphenyl)benzenepropanamide
CAS 393121-74-9
Quantity 25 mg
Molecular Formula C16H17NO2
Formula Weight 255.31
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.