Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AACOCF3
GSA_VAGREENER_CHOICE

Catalog No. p-4783216
Click to view available options
Quantity:
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Phospholipase A2 inhibitor

Inhibitor of cytosolic (85 kDa) phospholipase A2. Also inhibits fatty acid amide hydrolase (FAAH, anandamide amidase) in vitro.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 149301-79-1
Molecular Formula C21H31F3O

Specifications

Quantity 5 mg
Formula Weight 356.47
Purity >98%
Molecular Weight (g/mol) 356.47
Chemical Name or Material AACOCF3
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.