Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AADACL1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP276805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276805 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP276805 Supplier Novus Biologicals Supplier No. NBP276805
Add to Cart
Add to Cart

Goat Polyclonal Antibody

AADACL1 Polyclonal specifically detects AADACL1 in Human samples. It is validated for Flow Cytometry, Peptide ELISA, Immunofluorescence.

Specifications

Antigen AADACL1
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 0.50 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 10 ug/ml, Peptide ELISA Detection Limit 1:1000, Immunofluorescence 10 ug/ml
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_065843.3, NP_001139748.2, NP_001139749.1
Gene Alias AADACL1, EC 3.1.1.79, KIAA1363EC 3.1.1, NCEHArylacetamide deacetylase-like 1EC 3.1.1.-, neutral cholesterol ester hydrolase 1
Gene Symbols NCEH1
Host Species Goat
Immunogen Peptide with sequence C-RTRNSYIKWLDQN, from the C Terminus of the protein sequence according to NP_065843.3; NP_001139748.2; NP_001139749.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Primary or Secondary Primary
Gene ID (Entrez) 57552.0
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.