Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AAMDC Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen C11orf67
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP156786
SDP
View Documents
Novus Biologicals
NBP156786
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

AAMDC Polyclonal specifically detects AAMDC in Human samples. It is validated for Western Blot.
Specifications

Specifications

C11orf67
Polyclonal
Rabbit
Q9H7C9
28971
Synthetic peptides corresponding to C11ORF67 The peptide sequence was selected from the middle region of C11ORF67. Peptide sequence SEALKVPSSTVEYLKKHGIDVRVLQTEQAVKEYNALVAQGVRVGGVFHST.
Primary
Western Blot
Unconjugated
RUO
adipogenesis associated, Mth938 domain containing, C11orf67, chromosome 11 open reading frame 67, CK067, FLJ21035, hypothetical protein LOC28971, MGC3367, PTD015
AAMDC
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.