Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AAMDC Antibody (OTI1C4), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288618 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288618 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288618 Supplier Novus Biologicals Supplier No. NBP271840APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AAMDC Monoclonal antibody specifically detects AAMDC in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen AAMDC
Applications Western Blot
Classification Monoclonal
Clone OTI1C4
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias adipogenesis associated, Mth938 domain containing, C11orf67, chromosome 11 open reading frame 67, CK067, FLJ21035, hypothetical protein LOC28971, MGC3367, PTD015
Host Species Mouse
Immunogen Full length human recombinant protein of human C11orf67(NP_078960) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 28971
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.