Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AARSD1 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB065224 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB065224 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB065224 Supplier Novus Biologicals Supplier No. NBP300270IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AARSD1 Polyclonal specifically detects AARSD1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen AARSD1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias alanyl-tRNA editing protein Aarsd1, alanyl-tRNA synthetase domain containing 1, Alanyl-tRNA synthetase domain-containing protein 1, MGC2744
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant AARSD1 (Q9BTE6-2; Thr176-Glu525).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80755
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.