Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AASD-PPT Antibody, Novus Biologicals™
SDP

Catalog No. NBP154977 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP154977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP154977 Supplier Novus Biologicals Supplier No. NBP154977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AASD-PPT Polyclonal specifically detects AASD-PPT in Human samples. It is validated for Western Blot.

Specifications

Antigen AASD-PPT
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9NRN7
Gene Alias 4'-phosphopantetheinyl transferase, AASD-PPTDKFZp566E2346, Alpha-aminoadipic semialdehyde dehydrogenase-phosphopantetheinyl transferase, aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase, CGI-80, EC 2.7.8, EC 2.7.8.-, L-aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase, LYS2, LYS5, LYS5 ortholog
Gene Symbols AASDHPPT
Host Species Rabbit
Immunogen Synthetic peptides corresponding to AASD-PPT. The peptide sequence was selected from the C terminal of AASDHPPT. Peptide sequence SRHQDVPSQDDSKPTQRQFTILNFNDLMSSAVPMTPEDPSFWDCFCFTEE The peptide sequence for this immunogen was taken from within the described region.
Molecular Weight of Antigen 36 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 60496
Test Specificity Guinea pig: 85%; Bovine: 79%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.