Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AAV4 VP3 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB344964 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344964 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344964 Supplier Novus Biologicals Supplier No. NB10093576F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AAV4 VP3 Polyclonal antibody specifically detects AAV4 VP3 in Nonspecies specific, Virus samples. It is validated for Western Blot

Specifications

Antigen AAV4 VP3
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias Adenovirus 4, AdV 4
Host Species Rabbit
Immunogen Directed against a synthetic peptide within residues 450-475 of 81 kDa capsid VP3 protein of AAV 4.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific, Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.