Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AAV5 VP3 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB344959 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344959 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344959 Supplier Novus Biologicals Supplier No. NB10093577APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AAV5 VP3 Polyclonal antibody specifically detects AAV5 VP3 in Nonspecies specific, Virus samples. It is validated for Western Blot, Immunohistochemistry, In-situ Hybridization

Specifications

Antigen AAV5 VP3
Applications Western Blot, Immunohistochemistry, In Situ Hybridization (ISH)
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, In Situ Hybridization (ISH)
Formulation PBS
Gene Alias Adeno-Associated Virus, Adeno-Associated Virus (AAV)
Host Species Rabbit
Immunogen Synthetic peptide within residues 525-550 of 80 kDa capsid VP3 protein of AAV5.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Gene Therapy, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Non-species specific, Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.