Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AAV5 VP3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017926 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017926 0.025 mg
NB10093577 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017926 Supplier Novus Biologicals Supplier No. NB10093577SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AAV5 VP3 Polyclonal specifically detects AAV5 VP3 in Non-species specific, Virus samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen AAV5 VP3
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q9YIJ1
Gene Alias Adeno-Associated Virus, Adeno-Associated Virus (AAV)
Host Species Rabbit
Immunogen Synthetic peptide within residues 525-550 of 80 kDa capsid VP3 protein of AAV5.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Adeno-Associated Virus (AAV)
Target Species Virus, Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.