Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AbC™ Total Antibody Compensation Bead Kit
Click to view available options
Quantity:
100 tests
25 tests
Description
- Broadest multispecies reactivity—for use with all isotypes of mouse, rat, hamster, and rabbit antibody conjugates
- Brightest signal—most sensitive antibody-capture beads available
- Maximum laser compatibility—for use with all lasers
- Each kit provides polystyrene microspheres that have fluorescent antibody conjugate capture capacity (positive compensation beads) or are inert (negative compensation beads).
- Ideal for compensation with all isotypes of mouse (IgG1, IgG2a, IgG2b, IgG3), rat (IgG1, IgG2a, IgG2b), hamster (Armenian IgG and Syrian IgG), and rabbit (monoclonal and polyclonal) immunoglobulins.Demonstrates the highest and most consistent reactivity to the different subclasses of immunoglobulin in the industry.
Specifications
Specifications
| Diameter (Metric) | 6 μm |
| Content And Storage | Contains 1 vial AbC™ Total Compensation capture beads and 1 vial of negative beads. |
| Format | Tube |
| For Use With (Equipment) | Flow Cytometer |
| No. of Tests | 25 Tests |
| Quantity | 25 tests |
| Detection Method | Fluorescence |
| Excitation Wavelength Range | any lasers (UV to 633/635) |
| Shipping Condition | Room Temperature |
| Product Line | AbC |
| Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction