Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCA1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB400105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400105 0.1 mL
NB400105SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400105 Supplier Novus Biologicals Supplier No. NB400105
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 357 publications

ABCA1 Polyclonal specifically detects ABCA1 in Human, Mouse, Rat, Porcine, Canine, Chicken, Chinese Hamster, Equine, Hamster, Mustelid, Primate, Rabbit samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Gel Supershift Assay, Knockout Validated, Knockdown Validated, Block/Neutralize, PCR, Dual RNAscope ISH-IHC.

Specifications

Antigen ABCA1
Applications Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), KnockDown, Neutralization
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Simple Western 1:50, Chromatin Immunoprecipitation reported in scientific literature (PMID 19515742), Flow Cytometry 1:400. Use reported in scientific literature (PMID 21501868), ELISA reported in scientific literature (PMID 18541924), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100. Use reported in scientific literature (PMID 21501868), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 21846716; 21106520), Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen, Immunoblotting reported in scientific literature (PMID 27599291), Gel Super Shift Assays reported in scientific literature (PMID 15684432), Gel Supershift Assay, Chromatin Immunoprecipitation (ChIP), Knockout Validated reported in scientific literature (PMID 32273567), KnockDown Validated reported in scientific literature (PMID 31666189), Block/Neutralize reported in scientific literature (PMID 30821416), PCR reported in scientific literature (PMID 27406916), Dual RNAscope ISH-IHC
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. O95477
Gene Alias ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1
Gene Symbols ABCA1
Host Species Rabbit
Immunogen Partial peptide sequence (within residues 1100-1300) of human ABCA1 [UniProt# O95477]. Actual immunogen sequence is proprietary information.
Molecular Weight of Antigen 220 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Astrocyte Markers, Cancer, Cellular Markers, Cholesterol Metabolism, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 19
Test Specificity Human, hamster, mouse, rat, porcine, chicken and mustelid. Canine reactivity reported in scientific literature (PMID: 24612239).
Target Species Human, Mouse, Rat, Pig, Canine, Chicken, Chinese Hamster, Equine, Hamster, Mustelid, Primate, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.