Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCA1 Antibody (HJ1), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB1002068R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002068R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002068R Supplier Novus Biologicals Supplier No. NB1002068R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABCA1 Monoclonal specifically detects ABCA1 in Human, Mouse, Rat samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen ABCA1
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone HJ1
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Alias ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1
Gene Symbols ABCA1
Host Species Mouse
Immunogen 50 kDa N-terminal extracellular loop of mouse ABCA1 [UniProt# P41233]
Molecular Weight of Antigen 250 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Astrocyte Markers, Cancer, Cellular Markers, Cholesterol Metabolism, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 19
Target Species Human, Mouse, Rat, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.