Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCA4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$367.50 - $609.50
Specifications
Antigen | ABCA4 |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ABCA4 Polyclonal antibody specifically detects ABCA4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ABCA4 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
ABC Transporters, Lipid and Metabolism, Vision | |
PBS, pH 7.2, 40% glycerol | |
24 | |
IgG | |
Affinity purified |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
ABC10, ABCRRMP, ARMD2DKFZp781N1972, ATP binding cassette transporter, ATP-binding cassette sub-family A member 4, ATP-binding cassette transporter, retinal-specific, ATP-binding cassette, sub-family A (ABC1), member 4, ATP-binding transporter, retina-specific, CORD3, EC 3.6.3, FFMFLJ17534, photoreceptor rim protein, retinal-specific ATP-binding cassette transporter, retina-specific ABC transporter, RIM ABC transporter, RIM protein, RmP, RP19, Stargardt disease protein, STGD, STGD1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: IQSQRKGSEGTCSCSSKGFSTTCPAHVDDLTPEQVLDGDVNELMDVVLHHVPEAKLVECIGQELIFLLPNKNFKHRAYASLFRELEET | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title