Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCA5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15980920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15980920 20 μL
NBP159809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15980920 Supplier Novus Biologicals Supplier No. NBP15980920UL

Rabbit Polyclonal Antibody

ABCA5 Polyclonal specifically detects ABCA5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ABCA5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8WWZ7
Gene Alias ABC13, ATP-binding cassette A5, ATP-binding cassette sub-family A member 5, ATP-binding cassette, sub-family A (ABC1), member 5, DKFZp451F117, DKFZp779N2435, EC 3.6.3, EC 3.6.3.25, EC 3.6.3.41, EST90625, FLJ16381, KIAA1888
Gene Symbols ABCA5
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ABCA5(ATP-binding cassette, sub-family A (ABC1), member 5) The peptide sequence was selected from the C terminal of ABCA5. Peptide sequence HKEYDDKKDFLLSRKVKKVATKYISFCVKKGEILGLLGPNGAGKSTIINI.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 23461
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.