Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCA7 Antibody (7A1-144), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB400163V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400163V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400163V Supplier Novus Biologicals Supplier No. NB400163V
Add to Cart
Add to Cart

Rat Monoclonal Antibody

ABCA7 Monoclonal specifically detects ABCA7 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ABCA7
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7A1-144
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin
Gene Alias ABCA-SSN, ABCXATP-binding cassette sub-family A member 7, ATP-binding cassette, sub-family A (ABC1), member 7, Autoantigen SS-N, EC 2.2.1.1, EC 3.6.3, EC 3.6.3.41, FLJ40025, Macrophage ABC transporter
Gene Symbols ABCA7
Host Species Rat
Immunogen Mouse ABCA7 stably-transfected HeLa cells. [UniProt# Q91V24]
Molecular Weight of Antigen 240 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Astrocyte Markers, Cellular Markers, Cholesterol Metabolism, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10347
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.