Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCA8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP120867 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120867 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP120867 Supplier Novus Biologicals Supplier No. NBP120867
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ABCA8 Polyclonal specifically detects ABCA8 in Human samples. It is validated for Peptide ELISA.

Specifications

Antigen ABCA8
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:4000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_009099.1
Gene Alias ATP-binding cassette sub-family A member 8, ATP-binding cassette, sub-family A (ABC1), member 8, EC 3.6.3, EC 3.6.3.41, KIAA0822MGC163152
Gene Symbols ABCA8
Host Species Goat
Immunogen Peptide with sequence C-KEQELGDFEEDFD corresponding to C-Terminus according to NP_009099.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10351
Test Specificity This antibody is expected to cross-react with the highly similar Human ABCA9, since the immunizing peptide differs from ABCA9 with only one residue (F8).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.