Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCA8 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167187 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167187 100 μg
NB167188 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167187 Supplier Novus Biologicals Supplier No. NBP316046100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABCA8 Polyclonal antibody specifically detects ABCA8 in Human, Rat samples. It is validated for Western Blot

Specifications

Antigen ABCA8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 50% glycerol, pH7.3
Gene Alias ATP-binding cassette sub-family A member 8, ATP-binding cassette, sub-family A (ABC1), member 8, EC 3.6.3, EC 3.6.3.41, KIAA0822MGC163152
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 425-495 of human ABCA8 (NP_009099.1). NEYGHRRPPLFFLKSSFWSQTQKTDHVALEDEMDADPSFHDSFEQAPPEFQGKEAIRIRNVTKEYKGKPDK
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10351
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.