Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCB5 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP177687G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177687G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177687G Supplier Novus Biologicals Supplier No. NBP177687G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABCB5 Polyclonal specifically detects ABCB5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ABCB5
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q2M3G0
Gene Alias ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5
Gene Symbols ABCB5
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human ABCB5 protein (between residues 150-200) [UniProt Q2M3G0]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 340273
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.