Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCB5 Antibody, Novus Biologicals™
SDP

Catalog No. NB10093365 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093365 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093365 Supplier Novus Biologicals Supplier No. NB10093365
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

ABCB5 Polyclonal specifically detects ABCB5 in Human samples. It is validated for Flow Cytometry, Peptide ELISA, Western Blot.

Specifications

Antigen ABCB5
Applications Flow Cytometry, Peptide ELISA, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/ml, Flow Cytometry 10 ug, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q2M3G0
Gene Alias ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5
Gene Symbols ABCB5
Host Species Goat
Immunogen Peptide with sequence C-QTQHRNTSKKAQ corresponding to internal region according to NP_848654.3, NP_001157413.1.
Molecular Weight of Antigen 90 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline ABC Transporters, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 340273
Test Specificity This antibody is expected to recognize both reported isoforms (NP_848654.3; NP_001157413.1)
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.