Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCB7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP221600 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221600 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221600 Supplier Novus Biologicals Supplier No. NBP221600
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABCB7 Polyclonal antibody specifically detects ABCB7 in Human, Mouse, Drosophila samples. It is validated for Western Blot.

Specifications

Antigen ABCB7
Applications Western Blot
Classification Polyclonal
Concentration 0.6mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS pH7, 20% Glycerol with 0.025% Proclin 300
Gene Accession No. O75027
Gene Alias ABC transporter 7 protein, ABC7ATP-binding cassette sub-family B member 7, mitochondrial, ASATATP-binding cassette 7, Atm1p, ATP-binding cassette transporter 7, ATP-binding cassette, sub-family B (MDR/TAP), member 7, EC 3.6.3, EC 3.6.3.42, EST140535
Gene Symbols ABCB7
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ABCB7. The exact sequence is proprietary.
Molecular Weight of Antigen 83 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 22
Test Specificity Human. Expected to cross react based on sequence identity: Mouse (97%), Rat (98%), Bovine (97%).
Target Species Human, Mouse, Drosophila
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.