Learn More
ABCB8 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $649.44
Specifications
| Antigen | ABCB8 |
|---|---|
| Concentration | 80 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0150
![]()
|
Rockland Immunochemicals
100401P00S |
25 μL |
Each for $178.35
|
|
|||||
50-199-0149
![]()
|
Rockland Immunochemicals
100401P00 |
100 μL |
Each for $649.44
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. A BLAST analysis was used to suggest cross-reactivity with ABCB8 from human, mouse, and rat based on a 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known. The antibody detects ABCB8 in cell lysates.
This nuclear gene encodes a multi-pass membrane protein that is targeted to the mitochondrial inner membrane. The encoded protein is an ATP-dependent transporter that may mediate the passage of organic and inorganic molecules out of the mitochondria. Loss of function of the related gene in mouse results in a disruption of iron homeostasis between the mitochondria and cytosol. Alternative splicing results in multiple transcript variants.Specifications
| ABCB8 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9NUT2 | |
| 11194 | |
| ABCB8 was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a internal portion of human ABCB8. | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| ABCB8 |
| 80 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| 4833412N02Rik; AA409895; ABC transporter 8; Abcb8; ATP binding cassette subfamily B member 8; ATP-binding cassette sub-family B member 8, mitochondrial; ATP-binding cassette, subfamily B (MDR/TAP), member 8; ATP-binding cassette, sub-family B (MDR/TAP), member 8; ATP-binding cassette, sub-family B, member 8; BOS_4761; EST328128; hypothetical protein LOC548340; MABC1; M-ABC1; mitochondrial ABC protein; Mitochondrial ATP-binding cassette 1; zgc:113037 | |
| ABCB8 | |
| Primary | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.