Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCC13 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146143 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146143 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146143 Supplier Novus Biologicals Supplier No. NBP146143
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ABCC13 Polyclonal specifically detects ABCC13 in Human, Canine samples. It is validated for Peptide ELISA.

Specifications

Antigen ABCC13
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. AAL47201.1
Gene Alias ATP-binding cassette protein C13, ATP-binding cassette, sub-family C (CFTR/MRP), member 13, pseudogene, C21orf73PRED6ATP-binding cassette, sub-family C (CFTR/MRP), member 13
Gene Symbols ABCC13
Host Species Goat
Immunogen Peptide with sequence CEHSSDFGWNGY corresponding to internal region according to AAL47201.1, AAQ22368.1, EAX10059.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150000
Test Specificity This antibody is expected to recognize all reported isoforms (AAL47201.1; AAQ22368.1; EAX10059.1).
Target Species Human, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.