Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCD1 Antibody (OTI2C12), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP27010A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27010A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27010A30 Supplier Novus Biologicals Supplier No. NBP270150AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABCD1 Monoclonal specifically detects ABCD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated, Knockdown Validated.

Specifications

Antigen ABCD1
Applications Western Blot, Immunohistochemistry, Immunoassay, KnockDown
Classification Monoclonal
Clone OTI2C12
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Knockout Validated, Knockdown Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Adrenoleukodystrophy protein, ALDATP-binding cassette sub-family D member 1, ALDPadrenoleukodystrophy, AMNABC42, ATP-binding cassette, sub-family D (ALD), member 1
Gene Symbols ABCD1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 508-745 of human ABCD1 (NP_000024) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline ABC Transporters, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 215.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.