Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCE1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB40116A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB40116A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB40116A350 Supplier Novus Biologicals Supplier No. NB400116AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABCE1 Polyclonal specifically detects ABCE1 in Human, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ABCE1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 2′-5′-oligoadenylate-binding protein, ATP-binding cassette, sub-family E (OABP), member 1, HuHP68, OABP, Ribonuclease 4 inhibitor, ribonuclease L (2′-5′-oligoisoadenylate synthetase-dependent) inhibitor, RLIABC38, RNase L inhibitor, RNASEL1, RNASELIRNS4IATP-binding cassette sub-family E member 1
Gene Symbols ABCE1
Host Species Rabbit
Immunogen A fusion protein containing amino acids 1-119 of human ABCE1 [UniProtNo. P61221]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline ABC Transporters, Immune System Diseases, Immunology, Lipid and Metabolism, Membrane Trafficking and Chaperones, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6059.0
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.