Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCF1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP118889 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP118889 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP118889 Supplier Novus Biologicals Supplier No. NBP118889
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABCF1 Polyclonal specifically detects ABCF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative).

Specifications

Antigen ABCF1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoprecipitation (Negative)
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_001020262.1
Gene Alias ABC27, ABC50TNFalpha-inducible ATP-binding protein, ATP-binding cassette 50, ATP-binding cassette 50 (TNF-alpha stimulated), ATP-binding cassette sub-family F member 1, ATP-binding cassette, sub-family F (GCN20), member 1, EC 2.7.7.6, EC 3.6.3.17, EST123147, TNF-alpha-stimulated ABC protein
Gene Symbols ABCF1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 795 and 845 of human ATP-binding cassette 50 using the numbering given in entry NP_001020262.1 (GeneID 23).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.