Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCF1 Rabbit anti-Human, Clone: SN07-12, Novus Biologicals™
SDP

Catalog No. NBP267698 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267698 Supplier Novus Biologicals Supplier No. NBP267698

Rabbit Monoclonal Antibody

ABCF1 Monoclonal specifically detects ABCF1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ABCF1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SN07-12
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:400
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias ABC27, ABC50TNFalpha-inducible ATP-binding protein, ATP-binding cassette 50, ATP-binding cassette 50 (TNF-alpha stimulated), ATP-binding cassette sub-family F member 1, ATP-binding cassette, sub-family F (GCN20), member 1, EC 2.7.7.6, EC 3.6.3.17, EST123147, TNF-alpha-stimulated ABC protein
Gene Symbols ABCF1
Host Species Rabbit
Immunogen Synthetic peptide within Human ABCF1 aa 796-845/ 845. (SwissProt: Q8NE71 Human; SwissProt: Q6MG08 Rat)
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 23
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.