Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCF2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB107055 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB107055 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB107055 Supplier Novus Biologicals Supplier No. NBP298977AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABCF2 Polyclonal specifically detects ABCF2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ABCF2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias ABC28, ABC-type transport protein, ATP-binding cassette sub-family F member 2, ATP-binding cassette, sub-family F (GCN20), member 2, DKFZp586K1823, EC 3.6.3, EC 3.6.3.17, EST133090, HUSSY18, HUSSY-18, Iron inhibited ABC transporter 2, Iron-inhibited ABC transporter 2, M-ABC1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human ABCF2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Chromatin Research, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, Membrane Trafficking and Chaperones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10061
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.