Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCG1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB400132UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400132UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400132UV Supplier Novus Biologicals Supplier No. NB400132UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABCG1 Polyclonal specifically detects ABCG1 in Human, Mouse, Rat, Chinese Hamster, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Flow (Intracellular).

Specifications

Antigen ABCG1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Flow (Intracellular)
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ABC transporter 8, ABC8ATP-binding cassette sub-family G member 1, ATP-binding cassette transporter 8, ATP-binding cassette, sub-family G (WHITE), member 1, homolog of Drosophila white, MGC34313, White protein homolog, white protein homolog (ATP-binding cassette transporter 8), 10ATP-binding cassette transporter member 1 of subfamily G, WHITE1, WHT1
Gene Symbols ABCG1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of human ABCG1 (between residues 300-400). [UniProtNo. P45844]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline ABC Transporters, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9619.0
Target Species Human, Mouse, Rat, Chinese Hamster, Hamster, Monkey, Rabbit
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.