Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCG2/CD338 Antibody (MM0047-2J39) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018045 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018045 0.025 mg
NB110935111 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018045 Supplier Novus Biologicals Supplier No. NB110935110.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABCG2/CD338 Monoclonal specifically detects ABCG2/CD338 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen ABCG2/CD338
Applications CyTOF, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Monoclonal
Clone MM0047-2J39
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 1:100-1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1000, CyTOF-ready
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Alias ABC transporter, ABC15, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter
Gene Symbols ABCG2
Host Species Mouse
Immunogen human recombinant rHABCG2 protein
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline ABC Transporters, Cancer, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 9429
Test Specificity This was selected for its ability to detect human ABCG2 by flow cytometry and immunocytochemistry.
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.