Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCG2/CD338 Antibody (MM0047-2J39) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $564.00
Specifications
| Antigen | ABCG2/CD338 |
|---|---|
| Clone | MM0047-2J39 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 1:100-1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1000, CyTOF-ready |
| Applications | CyTOF, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018045
![]() |
Novus Biologicals
NB110935110.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB110935111
![]() |
Novus Biologicals
NB110935110.1MG |
0.1 mg |
Each for $564.00
|
|
|||||
Description
ABCG2/CD338 Monoclonal specifically detects ABCG2/CD338 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| ABCG2/CD338 | |
| LYOPH | |
| CyTOF, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| ABC Transporters, Cancer, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Stem Cells | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| 9429 | |
| human recombinant rHABCG2 protein | |
| Primary | |
| Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
| This was selected for its ability to detect human ABCG2 by flow cytometry and immunocytochemistry. |
| MM0047-2J39 | |
| Flow Cytometry 1:100-1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ABC transporter, ABC15, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter | |
| ABCG2 | |
| IgG2 | |
| Protein G purified | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title