Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCG8 Antibody (1B10A5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP171706A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP171706A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP171706A94 Supplier Novus Biologicals Supplier No. NBP171706AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABCG8 Monoclonal specifically detects ABCG8 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen ABCG8
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Classification Monoclonal
Clone 1B10A5
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ATP-binding cassette sub-family G member 8, ATP-binding cassette, sub-family G (WHITE), member 8, ATP-binding cassette, sub-family G (WHITE), member 8 (sterolin 2), GBD4ATP-binding cassette, subfamily G, member 8, MGC142217, sterolin 2, sterolin-2, STSL
Gene Symbols ABCG8
Host Species Mouse
Immunogen Full length mouse ABCG8 [Swiss-ProtNo. Q9DBM0]
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline ABC Transporters, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64241.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.