Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCG8 Antibody (1B10A5), Novus Biologicals™
SDP

Catalog No. NBP171706 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP171706 0.1 mL
NBP171706SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP171706 Supplier Novus Biologicals Supplier No. NBP171706
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 8 publications

ABCG8 Monoclonal specifically detects ABCG8 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen ABCG8
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay
Classification Monoclonal
Clone 1B10A5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 25 μg, Immunohistochemistry-Paraffin 1:100, Knockout Validated
Gene Accession No. Q9DBM0
Gene Alias ATP-binding cassette sub-family G member 8, ATP-binding cassette, sub-family G (WHITE), member 8, ATP-binding cassette, sub-family G (WHITE), member 8 (sterolin 2), GBD4ATP-binding cassette, subfamily G, member 8, MGC142217, sterolin 2, sterolin-2, STSL
Gene Symbols ABCG8
Host Species Mouse
Immunogen Full length mouse ABCG8 [Swiss-Prot# Q9DBM0]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64241
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.