Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCG8 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB400117H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400117H 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400117H Supplier Novus Biologicals Supplier No. NB400117H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABCG8 Polyclonal specifically detects ABCG8 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen ABCG8
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Accession No. Q9H221
Gene Alias ATP-binding cassette sub-family G member 8, ATP-binding cassette, sub-family G (WHITE), member 8, ATP-binding cassette, sub-family G (WHITE), member 8 (sterolin 2), GBD4ATP-binding cassette, subfamily G, member 8, MGC142217, sterolin 2, sterolin-2, STSL
Gene Symbols ABCG8
Host Species Rabbit
Immunogen A synthetic peptide from the N-terminal region of human ABCG8 protein. [UniProt# Q9H221]
Molecular Weight of Antigen 70 kDa
Purification Method Affinity Purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64241
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.