Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abeta 40 Antibody, Novus Biologicals™
SDP

Catalog No. NBP144047 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144047 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144047 Supplier Novus Biologicals Supplier No. NBP144047
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

Abeta 40 Polyclonal specifically detects Abeta 40 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Abeta 40
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.12 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1:20000-1:60000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 50% (v/v) Glycerol with 1 mg/ml Bovine Serum Albumin (BSA) with No Preservative
Gene Accession No. P05067
Gene Alias amyloid beta 40, Beta-amyloid protein 40
Gene Symbols APP
Host Species Rabbit
Immunogen Abeta 40 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the C-terminus region of AB40. (Uniprot: P05067)
Molecular Weight of Antigen 4.33 kDa
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 351
Test Specificity A BLAST analysis was used to suggest reactivity with human, sheep, rabbit, canine, bovine, pig, guinea pig, and monkey based on 100% sequence homology. This antibody will not react with AB42. Cross-reactivity with Beta Amyloid from other sources has not been determined.
Reconstitution Reconstitute in 185μL of water if being used immediately.
Target Species Human, Mouse, Rat, Primate, Bovine, Canine, Guinea Pig, Monkey, Rabbit, Sheep
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.